Learn More
Research Products International Corp 2 Nitro BD Glucopyran 10G
SDP
Catalog No. 502230398
Encompass_Preferred
Spectrophotometric substrate for detection of β-D-Glucosidase.
Catalog No. 50-223-0398 Supplier Research Products International Corp Supplier No. N8102010.0
Spectrophotometric substrate for detection of β-D-Glucosidase.