Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitro-p-cresol, TCI America™
SDP

Catalog No. N0186500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0186500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0186500G Supplier TCI America Supplier No. N0186500G

missing translation for 'validMainframeMsg'

Specifications

CAS 119-33-5
Molecular Formula C7H7NO3
MDL Number MFCD00007120
UN Number 2446
Quantity 500 g
Synonym 2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
InChI Key SYDNSSSQVSOXTN-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)O)[N+](=O)[O-]
IUPAC Name 4-methyl-2-nitrophenol
Molecular Weight (g/mol) 153.137
PubChem CID 8391
Formula Weight 153.14
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Nitro-p-cresol
Show More Show Less
EINECSNumber (3)-0790
RTECSNumber GP2800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.