Learn More
Medchemexpress LLC 2-Nitroamino-2-imida 100g
SDP
Catalog No. 5000206349
Encompass_Preferred
2-Nitroamino-2-imidazoline is an intermediate in the synthesis of the neonicotinoid insecticide Imidacloprid (HY-B0838)[1]
Catalog No. 50-002-06349 Supplier Medchemexpress LLC Supplier No. HYW111370100G
2-Nitroamino-2-imidazoline is an intermediate in the synthesis of the neonicotinoid insecticide Imidacloprid (HY-B0838)[1]