Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitroanisole, 99%

Catalog No. AAA1951822
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1951822 100 g
AAA1951836 500 g
AAA195180E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1951822 Supplier Thermo Scientific Chemicals Supplier No. A1951822
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 91-23-6 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-23-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007096
InChI Key CFBYEGUGFPZCNF-UHFFFAOYSA-N
Synonym 2-nitroanisole, o-nitroanisole, anisole, o-nitro, 1-nitro-2-methoxybenzene, 2-methoxynitrobenzene, o-nitrophenyl methyl ether, benzene, 1-methoxy-2-nitro, 2-methoxy-1-nitrobenzene, benzene, methoxynitro, o-nitro methoxy benzene
PubChem CID 7048
ChEBI CHEBI:48722
IUPAC Name 1-methoxy-2-nitrobenzene
SMILES COC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 9°C to 10°C
Density 1.254
Boiling Point 272°C to 273°C
Flash Point 142°C
Refractive Index 1.561
Quantity 100 g
UN Number UN2730
Beilstein 1868032
Merck Index 14,6585
Formula Weight 153.14
Percent Purity 99%
Chemical Name or Material 2-Nitroanisole
Show More Show Less
Hazard Category H302-H350
Hazard Statement GHS H Statement

H350-H302

May cause cancer.

Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROANISOLE, LIQUID
EINECSNumber 202-052-1
RTECSNumber BZ8790000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.