Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Nitrobenzaldehyde

Catalog No. N108022G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
2 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N108022G 2 g
N10802100G 100 g
N1080225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N108022G Supplier Sigma Aldrich Supplier No. N108022G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Nitrobenzaldehyde

Specifications

CAS 552-89-6
Melting Point 42°C to 44°C (lit.)
Boiling Point 153°C (23 mmHg)
Molecular Formula C7H5NO3
Linear Formula O2NC6H4CHO
MDL Number MFCD00007132
Quantity 2 g
Synonym o-NBA; ortho-nitrobenzaldehyde
Molecular Weight (g/mol) 151.12
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.