Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Nitrobenzaldehyde | 552-89-6 | 151.12 | 100 G
SDP

Catalog No. 5000460410
Encompass_Preferred

2-Nitrobenzaldehyde is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • CAS number: 552-89-6
  • Molecular formula: C7H5NO3
  • Molecular weight: 151.12
  • Purity: 99.99%
  • Appearance: Solid, off-white to light yellow
  • Storage conditions: 4°C under nitrogen; -80°C (6 months) or -20°C (1 month) in solvent under nitrogen
  • Solubility: 100 mg/mL in DMSO with warming

Catalog No. 50-004-60410 Supplier Medchemexpress LLC Supplier No. HYW007454100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Nitrobenzaldehyde is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • CAS number: 552-89-6
  • Molecular formula: C7H5NO3
  • Molecular weight: 151.12
  • Purity: 99.99%
  • Appearance: Solid, off-white to light yellow
  • Storage conditions: 4°C under nitrogen; -80°C (6 months) or -20°C (1 month) in solvent under nitrogen
  • Solubility: 100 mg/mL in DMSO with warming

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.