Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Nitrobenzaldehyde | 552-89-6 | 151.12 | 250 G
SDP

Catalog No. 5000449961
Encompass_Preferred

2-Nitrobenzaldehyde is a biochemical reagent utilized as a biological material or organic compound for various life science research applications.

  • Biochemical reagent for life science research
  • Can be used as a biological material or organic compound
  • Appears as an off-white to light yellow solid
  • Recommended storage at 4°C under nitrogen
  • For solutions, store at -80°C for up to 6 months or -20°C for 1 month under nitrogen
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-49961 Supplier Medchemexpress LLC Supplier No. 250GHYW007454
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Nitrobenzaldehyde is a biochemical reagent utilized as a biological material or organic compound for various life science research applications.

  • Biochemical reagent for life science research
  • Can be used as a biological material or organic compound
  • Appears as an off-white to light yellow solid
  • Recommended storage at 4°C under nitrogen
  • For solutions, store at -80°C for up to 6 months or -20°C for 1 month under nitrogen
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.