Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Nitrobenzaldehyde | 552-89-6 | 151.12 | 50 G
SDP

Catalog No. 5000460305
Encompass_Preferred

2-Nitrobenzaldehyde is a biochemical reagent used as a biological material or organic compound for life science-related research.

  • Molecular weight: 151.12
  • Molecular formula: C7H5NO3
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Storage: 4°C, under nitrogen; in solvent: -80°C for 6 months or -20°C for 1 month, under nitrogen
  • Target: Biochemical assay reagents
  • In vitro solubility: 100 mg/mL in DMSO (ultrasonic, warming to 60°C)
  • In vivo solubility recommendations are available with various solvent mixtures

Catalog No. 50-004-60305 Supplier Medchemexpress LLC Supplier No. HYW00745450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Nitrobenzaldehyde is a biochemical reagent used as a biological material or organic compound for life science-related research.

  • Molecular weight: 151.12
  • Molecular formula: C7H5NO3
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Storage: 4°C, under nitrogen; in solvent: -80°C for 6 months or -20°C for 1 month, under nitrogen
  • Target: Biochemical assay reagents
  • In vitro solubility: 100 mg/mL in DMSO (ultrasonic, warming to 60°C)
  • In vivo solubility recommendations are available with various solvent mixtures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.