Learn More
Medchemexpress LLC 2-nitrobenzaldehyde semicarbazone-13C,15N2 | 760179-80-4 | 98.0% | 211.15 g/mol | C7¹³CH8N2¹⁵N2O3 | 5 MG
SDP
Catalog No. 5000262654
Encompass_Preferred
2-Nitrobenzaldehyde semicarbazone-13C,15N2 is an isotopically labeled analytical standard used as an internal standard or tracer for LC-MS/MS quantitation of semicarbazide. The compound contains 13C and two 15N atoms to produce a reliable mass shift for MS detection and is supplied in milligram-scale quantities for analytical workflows.
  • Isotopically labeled with 13C and 15N2 for a clear mass shift in MS analyses.
  • High purity (≈98.0%) suitable for quantitative LC-MS/MS applications.
  • Molecular weight 211.15 g/mol compatible with standard MS methods.
  • Supplied in small mg-scale vials for tracer and standardization studies.
  • Available documentation includes datasheet, COA, and SDS to support analytical use.
Catalog No. 50-002-62654 Supplier Medchemexpress LLC Supplier No. HY133706S5MG
2-Nitrobenzaldehyde semicarbazone-13C,15N2 is an isotopically labeled analytical standard used as an internal standard or tracer for LC-MS/MS quantitation of semicarbazide. The compound contains 13C and two 15N atoms to produce a reliable mass shift for MS detection and is supplied in milligram-scale quantities for analytical workflows.
  • Isotopically labeled with 13C and 15N2 for a clear mass shift in MS analyses.
  • High purity (≈98.0%) suitable for quantitative LC-MS/MS applications.
  • Molecular weight 211.15 g/mol compatible with standard MS methods.
  • Supplied in small mg-scale vials for tracer and standardization studies.
  • Available documentation includes datasheet, COA, and SDS to support analytical use.