Learn More
Medchemexpress LLC 2-Nitrobenzenesulfonamide | 5455-59-4 | 202.19 | 100 G

Supplier: Medchemexpress LLC HYW008096100G
2-Nitrobenzenesulfonamide is a biochemical reagent suitable for life science research. This organic compound is intended for research use only and is not for patient application.
- Purity: 99.96%
- Molecular weight: 202.19
- Formula: C6H6N2O4S
- CAS number: 5455-59-4
- Appearance: Solid
- Color: White to yellow
- Ships at room temperature in continental US
- Powder storage: -20°C for 3 years or 4°C for 2 years
- Storage in solvent: -80°C for 6 months or -20°C for 1 month
- Related classifications: Biochemical Assay Reagents
- Includes data sheet, COA, SDS, HNMR, and RP-HPLC documentation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.