Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzoic acid, 95%

Catalog No. AC278320050
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
1 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC278320050 5 g Glass bottle
AC278320250 25 g Glass bottle
AC278322500 250 g Glass bottle
AC278320010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC278320050 Supplier Thermo Scientific Chemicals Supplier No. 278320050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 552-16-9 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-16-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007137
InChI Key SLAMLWHELXOEJZ-UHFFFAOYSA-N
Synonym 2-nitrobenzoic acid, o-nitrobenzoic acid, benzoic acid, 2-nitro, 2-nitrobenzoicacid, o-carboxynitrobenzene, benzoic acid, o-nitro, nitrobenzoic acid, 2-nitro-benzoic acid, ccris 2334, 2-nitro benzoic acid
PubChem CID 11087
ChEBI CHEBI:25620
SMILES C1=CC=C(C(=C1)C(=O)O)[N+](=O)[O-]

Specifications

CAS Min % 85.0
CAS Max % 100.0
Melting Point 141.0°C to 148.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4COOH
Quantity 5 g
Beilstein 09, 370
Fieser 13,231
Merck Index 14, 6589
Solubility Information Solubility in water: 6.8g/l. Other solubilities: 1g/3mL alcohol- 1g/2.5mL acetone,1g/220mL chloroform- 1g/2.5mL methanol,1g/4.5mL ether,slightly soluble in benzene,cs2,petroleum ether
Formula Weight 167.12
Percent Purity 95%
Physical Form Fine Powder
Chemical Name or Material 2-Nitrobenzoic acid, 85-90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-004-9
RTECSNumber DH5050000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.