Learn More
Medchemexpress LLC 2-nitrobenzyl alcohol | 612-25-9 | MFCD00007186 | 100.0% | 153.14 g/mol | C7H7NO3 | 500 G
SDP
Catalog No. 5000300010
Encompass_Preferred
2-nitrobenzyl alcohol is an aromatic nitro alcohol used as a reagent and synthetic building block in organic chemistry and biochemical research. It is commonly employed where a nitrobenzylic intermediate or photolabile precursor is required, and is supplied at high purity for laboratory research use only.
  • Aromatic nitro alcohol reagent for organic synthesis
  • High purity suitable for analytical and synthetic applications
  • Available in multiple pack sizes for scale flexibility
  • Solid form with typical handling and storage stability
  • Intended for research use only
Catalog No. 50-003-00010 Supplier Medchemexpress LLC Supplier No. HYW015569500G
2-nitrobenzyl alcohol is an aromatic nitro alcohol used as a reagent and synthetic building block in organic chemistry and biochemical research. It is commonly employed where a nitrobenzylic intermediate or photolabile precursor is required, and is supplied at high purity for laboratory research use only.
  • Aromatic nitro alcohol reagent for organic synthesis
  • High purity suitable for analytical and synthetic applications
  • Available in multiple pack sizes for scale flexibility
  • Solid form with typical handling and storage stability
  • Intended for research use only