Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrophenol, 98%

Catalog No. p-7082074
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1233330 250 g
AAA123330B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1233330 Supplier Thermo Scientific Chemicals Supplier No. A1233330
Only null left
Add to Cart

CAS: 88-75-5 | C6H5NO3 | 139.11 g/mol

2-Nitrophenol is a building block used as an acid-base indicator and corrosion inhibitor for aluminum-copper alloy. It is also used to make dyes, paint coloring, rubber chemicals, and substances that kill molds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrophenol is a building block used as an acid-base indicator and corrosion inhibitor for aluminum-copper alloy. It is also used to make dyes, paint coloring, rubber chemicals, and substances that kill molds.

Solubility
Soluble in water (2.1 g/L at 20°C), ethanol, chloroform, alcohol, and acetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-75-5
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00011688
InChI Key IQUPABOKLQSFBK-UHFFFAOYSA-N
Synonym o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
PubChem CID 6947
ChEBI CHEBI:16260
IUPAC Name 2-nitrophenol
SMILES OC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 44°C to 48°C
Density 1.26
Boiling Point 214°C to 216°C
Flash Point 108°C (226°F)
Odor Phenol-like
Quantity 250 g
UN Number UN1663
Beilstein 775403
Merck Index 14,6620
Solubility Information Soluble in water (2.1g/L at 20°C),ethanol,chloroform,alcohol,and acetone.
Formula Weight 139.11
Percent Purity 98%
Chemical Name or Material 2-Nitrophenol
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H373-H302
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROPHENOLS
EINECSNumber 201-857-5
RTECSNumber SM2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.