Learn More
Research Products International Corp 2-Nitrophenyl-B-D-Xylopyranoside, 500 Milligrams
SDP
Catalog No. 50213567
Encompass_Preferred
Spectrophotometric substrate for detection of β-Xylosidase.
Protect from moisture.
Catalog No. 50-213-567 Supplier Research Products International Corp Supplier No. N638000.5
Spectrophotometric substrate for detection of β-Xylosidase.
Protect from moisture.