Learn More
Research Products International Corp 2-Nitrophenyl-B-D-Glucopyranoside, 100 Milligrams
SDP
Catalog No. 50213577
Encompass_Preferred
Spectrophotometric substrate for detection of β-D-Glucosidase.
Catalog No. 50-213-577 Supplier Research Products International Corp Supplier No. N810200.1
Spectrophotometric substrate for detection of β-D-Glucosidase.