Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrophenyl n-Octyl Ether 98.0+%, TCI America™
SDP

Catalog No. S038010G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S038010G 10 g
S03801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S038010G Supplier TCI America Supplier No. S038010G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37682-29-4
Molecular Formula C14H21NO3
Molecular Weight (g/mol) 251.326
MDL Number MFCD00014693
InChI Key CXVOIIMJZFREMM-UHFFFAOYSA-N
Synonym 2-nitrophenyl octyl ether, 1-nitro-2-octyloxy benzene, octyl o-nitrophenyl ether, 2-nitrophenyl n-octyl ether, 2-octyloxy nitrobenzene, benzene, 1-nitro-2-octyloxy, 1-2-nitrophenoxy octane, 1-nitro-2-octyloxybenzene, 2-n-octyloxy nitrobenzene, npoe
PubChem CID 169952
IUPAC Name 1-nitro-2-octoxybenzene
SMILES CCCCCCCCOC1=CC=CC=C1[N+](=O)[O-]

Specifications

Boiling Point 198°C
Quantity 10 g
Formula Weight 251.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Nitrophenyl n-Octyl Ether [Matrix for FABMS and liquid SIMS]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.