Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Nitrophenyl octyl ether

Catalog No. 3651305G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-513-05G 5 g
36-513-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-513-05G Supplier Sigma Aldrich Supplier No. 3651305G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Nitrophenyl octyl ether (NPOE) is an organic solvent, which is mainly used as a plasticizer and a charge transferring liquid that can be used in electrochemical devices. Its water solubility is low and has a high molar volume with a relative permittivity of 24.2.NPOE can be used as a plasticizer in a polymeric inclusion membrane (PIM) that enhances the transport of copper(II) in the solution of ammonia. It can be incorporated in a cellulose tri-acetate/carbon nanotube (CTA/CNT) based membrane, which is used in the transport and detection of melamine from milk samples.

Specifications

CAS 37682-29-4
Density 1.04 g/mL (at 25°C)
Boiling Point 197°C to 198°C (11 mmHg)
Molecular Formula C14H21NO3
Linear Formula O2NC6H4O(CH2)7 CH3
Refractive Index n20/D 1.508 (literature)
Quantity 5 g
MDL Number MFCD00014693
Synonym 1-Nitro-2-octyloxybenzene
Molecular Weight (g/mol) 251.32
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.