Learn More
Research Products International Corp [ONPG] 2-Nitrophenyl-B-D-Galactopyranoside, 1 Gram
SDP
Catalog No. 50213574
Encompass_Preferred
Colorimetric and spectrophotometric substrate for detection of β-D-Galactosidase.
Catalog No. 50-213-574 Supplier Research Products International Corp Supplier No. N810001.0
Colorimetric and spectrophotometric substrate for detection of β-D-Galactosidase.