Learn More
Research Products International Corp [ONPG] 2-Nitrophenyl-B-D-Galactopyranoside, 5 Grams
SDP
Catalog No. 50213576
Encompass_Preferred
Colorimetric and spectrophotometric substrate for detection of β-D-Galactosidase.
Catalog No. 50-213-576 Supplier Research Products International Corp Supplier No. N810005.0
Colorimetric and spectrophotometric substrate for detection of β-D-Galactosidase.