Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Nitrophenylboronic acid | 5570-19-4 | 166.93 | 100 G
SDP

Catalog No. 5000447667
Encompass_Preferred

2-Nitrophenylboronic acid is a chemical compound commonly used as a laboratory chemical and in the manufacture of various substances. It has a molecular formula of C6H6BNO4 and a molecular weight of 166.93.

  • High purity: 99.99%
  • Appearance: White to light yellow solid
  • Suitable for laboratory use
  • Stable under recommended storage conditions
  • Long shelf life for powder at -20°C (3 years) or 4°C (2 years)
  • Storage in solvent for 6 months at -80°C or 1 month at -20°C

Catalog No. 50-004-47667 Supplier Medchemexpress LLC Supplier No. HYW002465100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Nitrophenylboronic acid is a chemical compound commonly used as a laboratory chemical and in the manufacture of various substances. It has a molecular formula of C6H6BNO4 and a molecular weight of 166.93.

  • High purity: 99.99%
  • Appearance: White to light yellow solid
  • Suitable for laboratory use
  • Stable under recommended storage conditions
  • Long shelf life for powder at -20°C (3 years) or 4°C (2 years)
  • Storage in solvent for 6 months at -80°C or 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.