Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Nitrophenylboronic acid | 5570-19-4 | 166.93 | 25 G
SDP

Catalog No. 5000449528
Encompass_Preferred

2-Nitrophenylboronic acid is a white to light yellow solid primarily for research use. This high-purity chemical is stable under recommended storage conditions.

  • White to light yellow solid
  • Purity of 99.99%
  • Stable under recommended storage conditions
  • Powder storage: 3 years at -20°C, 2 years at 4°C
  • Solution storage: 6 months at -80°C, 1 month at -20°C

Catalog No. 50-004-49528 Supplier Medchemexpress LLC Supplier No. 25GHYW002465
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Nitrophenylboronic acid is a white to light yellow solid primarily for research use. This high-purity chemical is stable under recommended storage conditions.

  • White to light yellow solid
  • Purity of 99.99%
  • Stable under recommended storage conditions
  • Powder storage: 3 years at -20°C, 2 years at 4°C
  • Solution storage: 6 months at -80°C, 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.