Learn More
Medchemexpress LLC HY-136590 10mg Medchemexpress, 2-Nitrosopyridine CAS:79917-37-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13659010MG
Encompass_Preferred
Medchemexpress, HY-136590 10mg 2-Nitrosopyridine CAS:79917-37-6 2-Nitrosopyridine is a nitroso compound that can be used to synthesize antibiotics. 2-Nitrosopyridine can be used as a Click or Diels-Alder derivatization reagent and an excellent dienophile. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1448