Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Nitrosotoluene

Catalog No. N264075G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N264075G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N264075G Supplier Sigma Aldrich Supplier No. N264075G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-methyl-1-indanone, a α-benzocycloalkenone, is a derivative of 1-indanone. its synthesis has been reported. the enzymatic dynamic kinetic resolution (dkr) of racemic 2-methyl-1-indanone has been studied. the asymmetric α-arylation and hydroxymethylation of 2-methyl-1-indanone has been reported. it participated in the synthesis of 2-methyl-6-carboxyazulene.2-methyl-1-indanone may be used as a starting material in the synthesis of α-benzocycloalkenone. it may be used in the synthesis of the following:
  • Cyclohex-2-en-1-yl 2-methyl-1h-inden-3-yl carbonate
  • 2-hydroxy-2-methyl-1-indanone
  • O-alkoxycarbonylation of lithium enolates

Specifications

CAS 611-23-4
Melting Point 72°C to 75°C (lit.)
Molecular Formula C7H7NO
Linear Formula CH3C6H4NO
MDL Number MFCD00002061
Quantity 25 g
Molecular Weight (g/mol) 121.14
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.