Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrotoluene, 99+%

Catalog No. AC129030025
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC129030025 2.5 L Glass bottle
AC129030010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC129030025 Supplier Thermo Scientific Chemicals Supplier No. 129030025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 88-72-2 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-72-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
InChI Key PLAZTCDQAHEYBI-UHFFFAOYSA-N
Synonym 2-nitrotoluene, o-nitrotoluene, o-nitrotoluol, o-methylnitrobenzene, benzene, 1-methyl-2-nitro, toluene, o-nitro, 2-methyl-1-nitrobenzene, 2-nitrotoluol, ortho-nitrotoluol, 2-methylnitrobenzene
PubChem CID 6944
ChEBI CHEBI:33098
IUPAC Name 1-methyl-2-nitrobenzene
SMILES CC1=CC=CC=C1[N+](=O)[O-]

Specifications

Melting Point -9.0°C
Density 1.1600g/mL
Boiling Point 222.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.5445 to 1.5465
Linear Formula CH3C6H4NO2
Quantity 2.5 L
Beilstein 05,318
Merck Index 15,6737
Specific Gravity 1.16
Solubility Information Solubility in water: 0.44g/L (20°C). Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 137.14
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Nitrotoluene
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 201-853-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.