Learn More
Cayman Chemical 2-Nonylquinolin-4(1H)-one, Purity: ≥95%, CAS: 55396-45-7, 50 mg
SDP
Catalog No. 502894541
Encompass_Preferred
A quinolone alkaloid; active against HCV (IC50 = 1.4 µg/ml) and L. gongylophorus; inhibits TPA- and A23187-induced NFAT activation in Jurkat cells but not LPS-induced NF-κB activation in RAW 264.7 cells (IC50s = 3.44 and >100 µM, respectively, in reporter assays)
Catalog No. 50-289-4541 Supplier Cayman Chemical Supplier No. 900362750 MG
A quinolone alkaloid; active against HCV (IC50 = 1.4 µg/ml) and L. gongylophorus; inhibits TPA- and A23187-induced NFAT activation in Jurkat cells but not LPS-induced NF-κB activation in RAW 264.7 cells (IC50s = 3.44 and >100 µM, respectively, in reporter assays)