Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Ntrophenyl beta-D-galac500MG
Catalog No. 502897456
Encompass
ONPG (2-Nitrophenyl beta-D-galactopyranoside) is a colorimetric substrate for beta-galactosidase.
Catalog No. 50-289-7456 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N1127500MG
ONPG (2-Nitrophenyl beta-D-galactopyranoside) is a colorimetric substrate for beta-galactosidase.