Learn More
Medchemexpress LLC 2'-O-(2-methoxyethyl)adenosine | 168427-74-5 | 99.0% | 325.32 g·mol⁻¹ | C13H19N5O5 | 1 ML

Supplier: Medchemexpress LLC HYW04849110MM/1ML
2′-O-(2-methoxyethyl)adenosine is an adenosine analogue and ATP derivative used as a building block in nucleotide and oligonucleotide chemistry. It is offered as a solid and as a ready-to-use 10 mM solution in DMSO (1 mL), and is supplied at high purity for synthetic applications.
- Available as solid (multiple mass options) and 10 mM solution in DMSO.
- High purity: 99.0%.
- Molecular weight 325.32 g·mol⁻¹; chemical formula C13H19N5O5.
- CAS number 168427-74-5.
- Storage: room temperature for continental US shipping; store at 4°C protected from light. In solvent: -80°C up to 6 months, -20°C up to 1 month (protect from light).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.