Learn More
Medchemexpress LLC 2'-O-(2-methoxyethyl)-cytidine | 223777-16-0 | 100.0% | 301.30 | C12H19N3O6 | 250 MG
SDP
Catalog No. 5000208886
Encompass_Preferred
2'-O-(2-Methoxyethyl)-cytidine is a 2'-O-modified cytidine nucleoside used as a building block in oligonucleotide synthesis and nucleic acid research. It is supplied as a purified reagent suitable for biochemical and molecular biology studies.
  • High purity (99.95%) for research use.
  • Modified nucleoside building block for oligonucleotide synthesis.
  • Supports studies of nucleic acid hybridization and stability.
  • Offered in a convenient 250 mg package for laboratory use.
Catalog No. 50-002-08886 Supplier Medchemexpress LLC Supplier No. 250MGHYW048496
2'-O-(2-Methoxyethyl)-cytidine is a 2'-O-modified cytidine nucleoside used as a building block in oligonucleotide synthesis and nucleic acid research. It is supplied as a purified reagent suitable for biochemical and molecular biology studies.
  • High purity (99.95%) for research use.
  • Modified nucleoside building block for oligonucleotide synthesis.
  • Supports studies of nucleic acid hybridization and stability.
  • Offered in a convenient 250 mg package for laboratory use.