Learn More
Medchemexpress LLC 2'-O-(2-Methoxyethyl)guanosine | 473278-54-5 | 99.8% | 341.32 | 1 ML
SDP
Catalog No. 5000402072
Encompass_Preferred
2′-O-(2-Methoxyethyl)guanosine (2'-O-MOE-rG) is a 2'-O-methoxyethyl-modified nucleoside analogue and an important intermediate in the synthesis of nucleic acid drugs. It is not effectively phosphorylated by cytosolic nucleoside kinases, nor is it incorporated into cellular DNA or RNA.
  • Important intermediate in the synthesis of nucleic acid drugs.
  • Not effectively phosphorylated by cytosolic nucleoside kinases.
  • Not incorporated into cellular DNA or RNA.
Catalog No. 50-004-02072 Supplier Medchemexpress LLC Supplier No. HY2378910MM/1ML
Add to Cart
Add to Cart
2′-O-(2-Methoxyethyl)guanosine (2'-O-MOE-rG) is a 2'-O-methoxyethyl-modified nucleoside analogue and an important intermediate in the synthesis of nucleic acid drugs. It is not effectively phosphorylated by cytosolic nucleoside kinases, nor is it incorporated into cellular DNA or RNA.
  • Important intermediate in the synthesis of nucleic acid drugs.
  • Not effectively phosphorylated by cytosolic nucleoside kinases.
  • Not incorporated into cellular DNA or RNA.