Learn More
Medchemexpress LLC 2'-O-(2-Methoxyethyl)guanosine | 473278-54-5 | 99.8% | 341.32 | 250 MG
SDP
Catalog No. 5000402402
Encompass_Preferred
2'-O-(2-Methoxyethyl)guanosine is a 2'-O-methoxyethyl-modified nucleoside analogue and an important intermediate in the synthesis of nucleic acid drugs. It is neither effectively phosphorylated by cytosolic nucleoside kinases nor incorporated into cellular DNA or RNA.
  • 2'-O-methoxyethyl-modified nucleoside analogue.
  • Important intermediate in nucleic acid drug synthesis.
  • Not effectively phosphorylated by cytosolic nucleoside kinases.
  • Not incorporated into cellular DNA or RNA.
  • Minimal effect on overall DNA/RNA synthesis in TK6 human lymphoblastoid cells at 10-1000 μM for 24 h.
Catalog No. 50-004-02402 Supplier Medchemexpress LLC Supplier No. 250MGHY23789
2'-O-(2-Methoxyethyl)guanosine is a 2'-O-methoxyethyl-modified nucleoside analogue and an important intermediate in the synthesis of nucleic acid drugs. It is neither effectively phosphorylated by cytosolic nucleoside kinases nor incorporated into cellular DNA or RNA.
  • 2'-O-methoxyethyl-modified nucleoside analogue.
  • Important intermediate in nucleic acid drug synthesis.
  • Not effectively phosphorylated by cytosolic nucleoside kinases.
  • Not incorporated into cellular DNA or RNA.
  • Minimal effect on overall DNA/RNA synthesis in TK6 human lymphoblastoid cells at 10-1000 μM for 24 h.