Learn More
Medchemexpress LLC 2'-O-(2-Methoxyethyl)guanosine | 473278-54-5 | 99.8% | 341.32 | 500 MG
SDP
Catalog No. 5000400926
Encompass_Preferred
2'-O-(2-Methoxyethyl)guanosine is a 2'-O-methoxyethyl-modified nucleoside analogue and an important intermediate in the synthesis of nucleic acid drugs. It is neither effectively phosphorylated by cytosolic nucleoside kinases, nor incorporated into cellular DNA or RNA. It is for research use only.
  • Nucleoside analogue
  • Important intermediate in nucleic acid drug synthesis
  • Not effectively phosphorylated by cytosolic nucleoside kinases
  • Not incorporated into cellular DNA or RNA
Catalog No. 50-004-00926 Supplier Medchemexpress LLC Supplier No. HY23789500MG
2'-O-(2-Methoxyethyl)guanosine is a 2'-O-methoxyethyl-modified nucleoside analogue and an important intermediate in the synthesis of nucleic acid drugs. It is neither effectively phosphorylated by cytosolic nucleoside kinases, nor incorporated into cellular DNA or RNA. It is for research use only.
  • Nucleoside analogue
  • Important intermediate in nucleic acid drug synthesis
  • Not effectively phosphorylated by cytosolic nucleoside kinases
  • Not incorporated into cellular DNA or RNA