Learn More
Medchemexpress LLC 2'-O-(2-Methoxyethyl) inosine | 545374-76-3 | C13H18N4O6 | 5 MG
SDP
Catalog No. 5000372367
Encompass_Preferred
2'-O-(2-Methoxyethyl) inosine is a purine nucleoside analog. Purine nucleoside analogs exhibit broad antitumor activity, particularly against indolent lymphoid malignancies. Its anticancer mechanisms involve the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analog
  • Exhibits broad antitumor activity, particularly against indolent lymphoid malignancies
  • Anticancer mechanisms involve the inhibition of DNA synthesis
  • Induces apoptosis
Catalog No. 50-003-72367 Supplier Medchemexpress LLC Supplier No. HY1525165MG
2'-O-(2-Methoxyethyl) inosine is a purine nucleoside analog. Purine nucleoside analogs exhibit broad antitumor activity, particularly against indolent lymphoid malignancies. Its anticancer mechanisms involve the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analog
  • Exhibits broad antitumor activity, particularly against indolent lymphoid malignancies
  • Anticancer mechanisms involve the inhibition of DNA synthesis
  • Induces apoptosis