Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-O-(2-methoxyethyl)-uridine | 223777-15-9 | 99.2% | 302.28 g·mol⁻¹ | C12H18N2O7 | 500 MG
SDP

Catalog No. 5000210306
Encompass_Preferred

2'-O-(2-methoxyethyl)-uridine is a chemically modified uridine nucleoside used as a research reagent and oligonucleotide building block. It is provided as a high-purity solid for laboratory research and method development and is intended for research use only; not for human use.

  • High purity (99.2%).
  • Molecular formula C12H18N2O7.
  • Molecular weight 302.28 g·mol⁻¹.
  • Soluble in DMSO (100 mg/mL); ultrasonic may be required.
  • Suitable as an oligonucleotide building block and research reagent.
  • Available in 500 MG and other package sizes.
  • Store protected from light; for solvent formulations, store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-10306 Supplier Medchemexpress LLC Supplier No. HYW048495500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-O-(2-methoxyethyl)-uridine is a chemically modified uridine nucleoside used as a research reagent and oligonucleotide building block. It is provided as a high-purity solid for laboratory research and method development and is intended for research use only; not for human use.

  • High purity (99.2%).
  • Molecular formula C12H18N2O7.
  • Molecular weight 302.28 g·mol⁻¹.
  • Soluble in DMSO (100 mg/mL); ultrasonic may be required.
  • Suitable as an oligonucleotide building block and research reagent.
  • Available in 500 MG and other package sizes.
  • Store protected from light; for solvent formulations, store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.