Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-O-(2-methoxyethyl)-uridine | 223777-15-9 | MFCD17215636 | 99.2% | 302.28 g/mol | C12H18N2O7 | 1 ML
SDP

Catalog No. 5000210342
Encompass_Preferred

2'-O-(2-methoxyethyl)-uridine is a chemically modified nucleoside derived from uridine and supplied for research applications. It is available as a 10 mM solution in DMSO (1 mL) and in solid quantities, and is characterized for purity and solubility to support nucleic acid chemistry and therapeutic development research.

  • Chemically modified uridine nucleoside.
  • Purity 99.2% as specified by supplier.
  • Available as 10 mM solution in DMSO (1 mL) and as solid quantities for experimental flexibility.
  • Soluble in DMSO; manufacturer lists in vivo formulation protocols and storage guidance.
  • Intended for research use only.

Catalog No. 50-002-10342 Supplier Medchemexpress LLC Supplier No. HYW04849510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-O-(2-methoxyethyl)-uridine is a chemically modified nucleoside derived from uridine and supplied for research applications. It is available as a 10 mM solution in DMSO (1 mL) and in solid quantities, and is characterized for purity and solubility to support nucleic acid chemistry and therapeutic development research.

  • Chemically modified uridine nucleoside.
  • Purity 99.2% as specified by supplier.
  • Available as 10 mM solution in DMSO (1 mL) and as solid quantities for experimental flexibility.
  • Soluble in DMSO; manufacturer lists in vivo formulation protocols and storage guidance.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.