Learn More
Medchemexpress LLC 2'-O,4'-C-Methyleneadenosine | 206055-70-1 | 99.9% | 279.26 | 250 MG
SDP
Catalog No. 5000347296
Encompass_Preferred
2'-O,4'-C-Methyleneadenosine (LNA-A) is a locked nucleic acid (LNA) and an adenosine analog. It is for research use only and not sold to patients. This product exhibits antiviral activity against wild type Hepatitis C virus genotype 1b in human HuH7 cells.
  • Locked nucleic acid (LNA)
  • Adenosine analog
  • High purity (99.9%)
  • Antiviral activity against wild type Hepatitis C virus genotype 1b in human HuH7 cells (EC50 > 50 μM)
Catalog No. 50-003-47296 Supplier Medchemexpress LLC Supplier No. 250MGHY110407
2'-O,4'-C-Methyleneadenosine (LNA-A) is a locked nucleic acid (LNA) and an adenosine analog. It is for research use only and not sold to patients. This product exhibits antiviral activity against wild type Hepatitis C virus genotype 1b in human HuH7 cells.
  • Locked nucleic acid (LNA)
  • Adenosine analog
  • High purity (99.9%)
  • Antiviral activity against wild type Hepatitis C virus genotype 1b in human HuH7 cells (EC50 > 50 μM)