Learn More
Cayman Chemical 2-O- a-D-Glucopyrnosylglyce 1g
SDP
Catalog No. 502946499
Encompass_Preferred
A compatible solute; prevents drying-induced membrane fusion and CF leakage in liposomes; levels are increased in response to osmotic stress in bacteria
Catalog No. 50-294-6499 Supplier Cayman Chemical Supplier No. 344921 G
A compatible solute; prevents drying-induced membrane fusion and CF leakage in liposomes; levels are increased in response to osmotic stress in bacteria