Learn More
Medchemexpress LLC 2-O-(alpha-D-glucopyranosyl)glycerol | 22160-26-5 | MFCD19980985 | 99.8% | 254.23 g/mol | C9H18O8 | 500 MG
SDP
Catalog No. 5000285223
Encompass_Preferred
2-O-(α-D-Glucopyranosyl)glycerol (glucosylglycerol) is a glycerol glycoside used as a biochemical research reagent. It acts as an osmolyte and bacterial metabolite and is commonly applied in studies of osmotic stress, microbial metabolism, and glycoside chemistry.
  • Functions as an osmolyte and bacterial metabolite.
  • Suitable for biochemical and microbiological research applications.
  • Water-soluble glycoside compatible with solution-based assays.
  • Commonly supplied as an aqueous solution for convenience in handling.
  • Well-characterized identifiers: CAS 22160-26-5; molecular weight 254.23 g/mol.
Catalog No. 50-002-85223 Supplier Medchemexpress LLC Supplier No. HYW353469500MG
2-O-(α-D-Glucopyranosyl)glycerol (glucosylglycerol) is a glycerol glycoside used as a biochemical research reagent. It acts as an osmolyte and bacterial metabolite and is commonly applied in studies of osmotic stress, microbial metabolism, and glycoside chemistry.
  • Functions as an osmolyte and bacterial metabolite.
  • Suitable for biochemical and microbiological research applications.
  • Water-soluble glycoside compatible with solution-based assays.
  • Commonly supplied as an aqueous solution for convenience in handling.
  • Well-characterized identifiers: CAS 22160-26-5; molecular weight 254.23 g/mol.