Learn More
Medchemexpress LLC 2"-O-beta-L-galactopyranosylorientin | 861691-37-4 | 99.9% | 610.52 | 10 MG
SDP
Catalog No. 5000365414
Encompass_Preferred
2"-O-beta-L-galactopyranosylorientin is extracted from the flowers of *Trollius ledebouri*. It is involved in transporter-mediated efflux in addition to passive diffusion and is a substrate of multidrug resistance protein 2 (MRP2). It also exhibits an anti-inflammatory effect.
  • Extracted from the flowers of *Trollius ledebouri*
  • Involves transporter-mediated efflux
  • Substrate of multidrug resistance protein 2 (MRP2)
  • Exhibits anti-inflammatory effect
Catalog No. 50-003-65414 Supplier Medchemexpress LLC Supplier No. 10MGHYN0406
2"-O-beta-L-galactopyranosylorientin is extracted from the flowers of *Trollius ledebouri*. It is involved in transporter-mediated efflux in addition to passive diffusion and is a substrate of multidrug resistance protein 2 (MRP2). It also exhibits an anti-inflammatory effect.
  • Extracted from the flowers of *Trollius ledebouri*
  • Involves transporter-mediated efflux
  • Substrate of multidrug resistance protein 2 (MRP2)
  • Exhibits anti-inflammatory effect