Learn More
Medchemexpress LLC 2-O-α-D-Glucopyranosyl-L-ascorbic Acid | 129499-78-1 | 100 MG
SDP
Catalog No. 5000419123
Encompass_Preferred
2-O-α-D-Glucopyranosyl-L-ascorbic Acid is an orally active glucoside derivative of ascorbic acid that can be hydrolyzed by α-glucosidase to release ascorbic acid. It is used in research related to tumors, inflammation, and other conditions.
  • Inhibits melanin synthesis
  • Prevents UV-induced cell damage
  • Promotes collagen synthesis in skin fibroblasts
  • Induces oxidative stress to inhibit tumor growth
Catalog No. 50-004-19123 Supplier Medchemexpress LLC Supplier No. HYN6821100MG
2-O-α-D-Glucopyranosyl-L-ascorbic Acid is an orally active glucoside derivative of ascorbic acid that can be hydrolyzed by α-glucosidase to release ascorbic acid. It is used in research related to tumors, inflammation, and other conditions.
  • Inhibits melanin synthesis
  • Prevents UV-induced cell damage
  • Promotes collagen synthesis in skin fibroblasts
  • Induces oxidative stress to inhibit tumor growth