Learn More
Medchemexpress LLC 2-O-β-D-Glucopyranosyl-L-ascorbic acid | 562043-82-7 | 10 MG
SDP
Catalog No. 5000420068
Encompass_Preferred
2-O-β-D-Glucopyranosyl-L-ascorbic acid (AA-2βG) is an orally active vitamin C derivative. It exhibits multiple activities including antioxidant, anti-tumor, and immunomodulatory effects. It mediates tumor cell apoptosis, induces cell cycle arrest, scavenges free radicals, and eliminates oxidative stress. It can be used in studies related to cancer, inflammation, and immunity.
  • Exhibits antioxidant, anti-tumor, and immunomodulatory effects.
  • Mediates tumor cell apoptosis and induces cell cycle arrest.
  • Scavenges free radicals and eliminates oxidative stress.
Catalog No. 50-004-20068 Supplier Medchemexpress LLC Supplier No. HYN695810MG
2-O-β-D-Glucopyranosyl-L-ascorbic acid (AA-2βG) is an orally active vitamin C derivative. It exhibits multiple activities including antioxidant, anti-tumor, and immunomodulatory effects. It mediates tumor cell apoptosis, induces cell cycle arrest, scavenges free radicals, and eliminates oxidative stress. It can be used in studies related to cancer, inflammation, and immunity.
  • Exhibits antioxidant, anti-tumor, and immunomodulatory effects.
  • Mediates tumor cell apoptosis and induces cell cycle arrest.
  • Scavenges free radicals and eliminates oxidative stress.