Learn More
Cayman Chemical 2-O-ethyl PAF C-16 10mg
SDP
Catalog No. 502894140
Encompass_Preferred
2-O-ethyl PAF C-16 is a synthetic PAF analog which contains an ethyl group, attached by an ether linkage, at the sn-2 position. It is a less potent agonist than methylcarbamyl PAF C-16 in the induction of platelet aggregation in both human and rabbit PRP.{613} 2-O-ethyl PAF C-16 causes aggregation of neutrophils in the presence of divalent cations and a desensitization to aggregation in the absence of divalent cations.{614}
Catalog No. 50-289-4140 Supplier Cayman Chemical Supplier No. 6092510 MG
2-O-ethyl PAF C-16 is a synthetic PAF analog which contains an ethyl group, attached by an ether linkage, at the sn-2 position. It is a less potent agonist than methylcarbamyl PAF C-16 in the induction of platelet aggregation in both human and rabbit PRP.{613} 2-O-ethyl PAF C-16 causes aggregation of neutrophils in the presence of divalent cations and a desensitization to aggregation in the absence of divalent cations.{614}