Learn More
Cayman Chemical 2-O-GaloylhyperIn 10mg
SDP
Catalog No. 502940123
Encompass_Preferred
A natural flavonoid; scavenges DPPH and ABTS radicals (IC50s = 3 and 3.5 μM, respectively); inhibits human OATP1B1 (IC50 = 19 μM in HEK293 cells); decreases LPS-induced IL-6 and TNF-α cytokine production from mouse RAW 264.7 macrophages at 15 μM; decreases LPS-induced mortality and liver damage in a mouse model of septic shock at 50 mg/kg
Catalog No. 50-294-0123 Supplier Cayman Chemical Supplier No. 2908610 MG
A natural flavonoid; scavenges DPPH and ABTS radicals (IC50s = 3 and 3.5 μM, respectively); inhibits human OATP1B1 (IC50 = 19 μM in HEK293 cells); decreases LPS-induced IL-6 and TNF-α cytokine production from mouse RAW 264.7 macrophages at 15 μM; decreases LPS-induced mortality and liver damage in a mouse model of septic shock at 50 mg/kg