Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-O-methoxyethyl-5-methyluridine | 163759-49-7 | MFCD02682964 | 99.8% | 316.31 g/mol | C13H20N2O7 | 250 MG
SDP

Catalog No. 5000208885
Encompass_Preferred

2'-O-methoxyethyl-5-methyluridine is a modified ribonucleoside used as a building block for oligonucleotide synthesis. It is supplied as a solid, high-purity material suitable for incorporation into modified RNA sequences to improve nuclease resistance and hybridization properties.

  • High purity (99.78%).
  • Molecular weight 316.31 g/mol.
  • Molecular formula C13H20N2O7.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Pack size 250 MG.
  • Store protected from light; in solution -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-08885 Supplier Medchemexpress LLC Supplier No. 250MGHYW048488
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-O-methoxyethyl-5-methyluridine is a modified ribonucleoside used as a building block for oligonucleotide synthesis. It is supplied as a solid, high-purity material suitable for incorporation into modified RNA sequences to improve nuclease resistance and hybridization properties.

  • High purity (99.78%).
  • Molecular weight 316.31 g/mol.
  • Molecular formula C13H20N2O7.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Pack size 250 MG.
  • Store protected from light; in solution -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.