Learn More
Medchemexpress LLC 2'-O-Methyl-4-thiouridine | 34218-80-9 | C10H14N2O5S | 25 MG
SDP
Catalog No. 5000374745
Encompass_Preferred
2'-O-Methyl-4-thiouridine is a modified uridine molecule, featuring a methoxy group at the 2' position of the ribose sugar and a thiol substitution at the 4-position of uridine. It is supplied in solid form with a purity of ≥95.0% and has a molecular weight of 274.29.
  • Utilized in the synthesis of nucleic acid molecules.
  • Serves as a modified sialic acid for messenger RNA (mRNA).
  • Enhances translation efficiency.
Catalog No. 50-003-74745 Supplier Medchemexpress LLC Supplier No. 25MGHY152513
2'-O-Methyl-4-thiouridine is a modified uridine molecule, featuring a methoxy group at the 2' position of the ribose sugar and a thiol substitution at the 4-position of uridine. It is supplied in solid form with a purity of ≥95.0% and has a molecular weight of 274.29.
  • Utilized in the synthesis of nucleic acid molecules.
  • Serves as a modified sialic acid for messenger RNA (mRNA).
  • Enhances translation efficiency.