Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-O-methyl-4-thiouridine | 34218-80-9 | MFCD30570246 | 99.4% | 274.29 g·mol-1 | C10H14N2O5S | 10 MG
SDP

Catalog No. 5000286336
Encompass_Preferred

2'-O-Methyl-4-thiouridine is a 2'-O-methylated thiouridine nucleoside analogue supplied as a solid for research use. It is used in biochemical and antitumor research and has reported activity related to inhibition of DNA synthesis and induction of apoptosis.

  • Purity approximately 99.4%.
  • CAS number 34218-80-9.
  • Molecular formula C10H14N2O5S; molecular weight 274.29 g·mol-1.
  • Appearance: solid, yellow to brown.
  • Solubility in water 5 mg/mL with ultrasonic assistance (<60°C).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solution -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical applications.

Catalog No. 50-002-86336 Supplier Medchemexpress LLC Supplier No. HY15251310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-O-Methyl-4-thiouridine is a 2'-O-methylated thiouridine nucleoside analogue supplied as a solid for research use. It is used in biochemical and antitumor research and has reported activity related to inhibition of DNA synthesis and induction of apoptosis.

  • Purity approximately 99.4%.
  • CAS number 34218-80-9.
  • Molecular formula C10H14N2O5S; molecular weight 274.29 g·mol-1.
  • Appearance: solid, yellow to brown.
  • Solubility in water 5 mg/mL with ultrasonic assistance (<60°C).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solution -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.