Learn More
Apexbio Technology LLC 2'-O-Methyl-5-methyl-UTP 10ul (100 mM)
SDP
Catalog No. 5000091993
Encompass_Preferred
2 -O-Methyl-5-methyl-UTP is a chemically modified nucleoside triphosphate analogue designed to enhance resistance to nuclease degradation and improve annealing properties toward complementary RNA sequences Incorporation of this compound into RNA oligonucleotides increases binding affinity to target RNA strands enhances stability under physiological conditions and reduces immune recognition Based on these pharmacological properties 2 -O-Methyl-5-methyl-UTP holds research potential in the synthesis of RNA therapeutics gene silencing projects antisense technology development and other applications requiring long-lasting bioactivity and minimal immunogenic response
Catalog No. 50-000-91993 Supplier Apexbio Technology LLC Supplier No. B79950.01
2 -O-Methyl-5-methyl-UTP is a chemically modified nucleoside triphosphate analogue designed to enhance resistance to nuclease degradation and improve annealing properties toward complementary RNA sequences Incorporation of this compound into RNA oligonucleotides increases binding affinity to target RNA strands enhances stability under physiological conditions and reduces immune recognition Based on these pharmacological properties 2 -O-Methyl-5-methyl-UTP holds research potential in the synthesis of RNA therapeutics gene silencing projects antisense technology development and other applications requiring long-lasting bioactivity and minimal immunogenic response