Learn More
Medchemexpress LLC 2'-O-methyl-5'-O-dmt-inosine-3'-CE-phosphoramidite | 128219-85-2 | MFCD08062007 | 98.0% | 784.84 | C41H49N6O8P | 250 MG
SDP
Catalog No. 5000285058
Encompass_Preferred
2'-O-Methyl-5'-O-dmt-inosine-3'-CE-phosphoramidite is a modified nucleoside phosphoramidite used for incorporation of inosine into oligonucleotides. It features a 2'-O-methyl ribose modification, a 5'-O-dimethoxytrityl protecting group, and a 3'-cyanoethyl phosphoramidite moiety compatible with automated solid-phase synthesis. Intended for research use, it supports generation of modified RNA and probes for biochemical studies.
  • Modified nucleoside for incorporation into oligonucleotides.
  • 2'-O-methyl modification increases nuclease resistance.
  • 5'-O-dimethoxytrityl protection enables stepwise synthesis monitoring.
  • 3'-cyanoethyl phosphoramidite suitable for automated solid-phase synthesis.
  • High purity suitable for research applications.
Catalog No. 50-002-85058 Supplier Medchemexpress LLC Supplier No. HYW392836250MG
2'-O-Methyl-5'-O-dmt-inosine-3'-CE-phosphoramidite is a modified nucleoside phosphoramidite used for incorporation of inosine into oligonucleotides. It features a 2'-O-methyl ribose modification, a 5'-O-dimethoxytrityl protecting group, and a 3'-cyanoethyl phosphoramidite moiety compatible with automated solid-phase synthesis. Intended for research use, it supports generation of modified RNA and probes for biochemical studies.
  • Modified nucleoside for incorporation into oligonucleotides.
  • 2'-O-methyl modification increases nuclease resistance.
  • 5'-O-dimethoxytrityl protection enables stepwise synthesis monitoring.
  • 3'-cyanoethyl phosphoramidite suitable for automated solid-phase synthesis.
  • High purity suitable for research applications.