Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-O-Methyl-8-methyl guanosine | 2306782-64-7 | 98.0% | C12H17N5O5 | 25 MG
SDP

Catalog No. 5000353071
Encompass_Preferred

2'-O-Methyl-8-methyl guanosine (m8Gm) is a Z-form RNA stabilizer. It can stabilize Z-RNA at low salt conditions, and m8Gm-contained oligonucleotides can stabilize Z-DNA under low salt conditions.

  • Stabilizes Z-RNA at low salt conditions
  • Oligonucleotides containing m8Gm can stabilize Z-DNA under low salt conditions
  • Molecular weight of 311.29
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-53071 Supplier Medchemexpress LLC Supplier No. 25MGHY145998
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-O-Methyl-8-methyl guanosine (m8Gm) is a Z-form RNA stabilizer. It can stabilize Z-RNA at low salt conditions, and m8Gm-contained oligonucleotides can stabilize Z-DNA under low salt conditions.

  • Stabilizes Z-RNA at low salt conditions
  • Oligonucleotides containing m8Gm can stabilize Z-DNA under low salt conditions
  • Molecular weight of 311.29
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.