Learn More
Apexbio Technology LLC 2'-O-Methyl-ATP(Synonyms: 2'-O-Methyladenosine 5'-triphosphate, 2'-OMe-ATP, ATP 2'-O-Methyl analog), 100ul (100 mM).
SDP
Catalog No. 5000060245
Encompass_Preferred
2 -O-Methyl-ATP is a chemically modified derivative of ATP in which the hydroxyl group at the ribose 2 position is replaced with a methyl group This ribose modification confers resistance to nuclease-mediated degradation thereby increasing its biological stability The modified nucleotide is commonly employed in biomedical research to investigate RNA structure-function relationships and RNA modification pathways and to study interactions influencing RNA polymerase specificity and catalytic activity Additionally 2 -O-Methyl-ATP serves as a tool in probe design for RNA interference experiments and aids in elucidating intracellular metabolic pathways involving nucleotide analogs
Catalog No. 50-000-60245 Supplier Apexbio Technology LLC Supplier No. B79680.1
2 -O-Methyl-ATP is a chemically modified derivative of ATP in which the hydroxyl group at the ribose 2 position is replaced with a methyl group This ribose modification confers resistance to nuclease-mediated degradation thereby increasing its biological stability The modified nucleotide is commonly employed in biomedical research to investigate RNA structure-function relationships and RNA modification pathways and to study interactions influencing RNA polymerase specificity and catalytic activity Additionally 2 -O-Methyl-ATP serves as a tool in probe design for RNA interference experiments and aids in elucidating intracellular metabolic pathways involving nucleotide analogs