Learn More
Apexbio Technology LLC 2'-O-Methyl-CTP(Synonyms: 2'-O-Methylcytidine-5'-triphosphate, 2'-OMe-CTP, 2'-O-Methyl CTP, 2'-Methoxy CTP), 10ul (100 mM).
SDP
Catalog No. 5000060261
Encompass_Preferred
2 -O-Methyl-CTP is a chemically modified analog of cytidine triphosphate (CTP) characterized by methyl substitution at the 2 -hydroxyl position of the ribose sugar This modification confers increased resistance against nuclease-mediated degradation when incorporated into RNA transcripts enhancing RNA stability It is commonly employed in biomedical research for investigating RNA structure-function relationships assessing RNA stability and synthesizing siRNA or miRNA analogs utilized in RNA interference (RNAi) studies
Catalog No. 50-000-60261 Supplier Apexbio Technology LLC Supplier No. B79690.01
2 -O-Methyl-CTP is a chemically modified analog of cytidine triphosphate (CTP) characterized by methyl substitution at the 2 -hydroxyl position of the ribose sugar This modification confers increased resistance against nuclease-mediated degradation when incorporated into RNA transcripts enhancing RNA stability It is commonly employed in biomedical research for investigating RNA structure-function relationships assessing RNA stability and synthesizing siRNA or miRNA analogs utilized in RNA interference (RNAi) studies